
4'-Nitro-3'-(trifluoromethyl)acetanilide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:393-12-4
:
Formula:C₉H₇F₃N₂O₃
MDL Number:MFCD00017994
:248.1600 g/mol
Melting Pt:123 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together