
2-Nitro-6-(trifluoromethyl)aniline 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:24821-17-8
:
Formula:C₇H₅F₃N₂O₂
MDL Number:MFCD08532494
:206.1220 g/mol
Melting Pt:48…52 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together