1-(4-Bromothiophen-2-yl)-2,2,2-trifluoroethanone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1252046-14-2
:
Formula:C₆H₂BrF₃OS
MDL Number:MFCD17168376
:259.05 g/mol
Storage Temperature:Ambient
Frequently Bought Together