Order Entry
Italy
ContactUsLinkComponent
2-(1,1-Difluoroethyl)-1,3-difluorobenzene 95%
2-(1,1-Difluoroethyl)-1,3-difluorobenzene 95%
Catalog # APOSPC520669-5G
Supplier:  Apollo Scientific
CAS Number:  
2-(1,1-Difluoroethyl)-1,3-difluorobenzene 95%
Catalog # APOSPC520669-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :178.13 g/mol
Storage Temperature:Ambient
CAS Number:1138445-06-3
Formula:C₈H₆F₄
MDL Number:MFCD11845808