1-(1,1-Difluoroethyl)-2,3-difluorobenzene 95%
Catalog # APOSPC520640-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:178.13 g/mol
Storage Temperature:Ambient
CAS Number:1138445-08-5
Formula:C₈H₆F₄
MDL Number:MFCD11845810