
6-(Trifluoromethyl)pyridine-3-sulphonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:959996-58-8
:
Formula:C₆H₃ClF₃NO₂S
MDL Number:MFCD10699112
:245.61 g/mol
Melting Pt:40…45 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together