Order Entry
Italy
ContactUsLinkComponent
2,6-Difluoro-3-(trifluoromethyl)pyridine 97%
Irritant
2,6-Difluoro-3-(trifluoromethyl)pyridine 97%
Catalog # APOSPC49654-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,6-Difluoro-3-(trifluoromethyl)pyridine 97%
Catalog # APOSPC49654-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :183.08 g/mol
Boiling Pt:70…71 °C/100mmHg
 :GHS07
Storage Temperature:Ambient
CAS Number:58584-98-8
Formula:C₆H₂F₅N
MDL Number:MFCD12546949