
Methyl-2-bromo-4,5-difluorobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:878207-28-4
:
Formula:C₈H₅BrF₂O₂
MDL Number:MFCD08458104
:251.03 g/mol
Melting Pt:28…29 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together