Ethyl-1-(4-phenyl-1,3-thiazol-2-yl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:159885-61-7
:
Formula:C₁₆H₁₂F₃N₃O₂S
MDL Number:MFCD00665285
:367.35 g/mol
Melting Pt:93…95 °C
Storage Temperature:Ambient
Frequently Bought Together