
Ethyl-4-methyl-2-[3-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:144061-14-3
:
Formula:C₁₄H₁₂F₃NO₂S
MDL Number:MFCD00142047
:315.32 g/mol
Melting Pt:91…93 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together