
2-(2,2,2-Trifluoroethoxy)ethanol 97+%
Catalog # APOSPC4692-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:144.09 g/mol
Boiling Pt:84 °C/80mmHg
Density:1.29 g/cm³ (25 °C)
Flash Pt:70 °C
:GHS07
Storage Temperature:Ambient
CAS Number:2358-54-5
Formula:C₄H₇F₃O₂
MDL Number:MFCD00462508