Order Entry
Italy
ContactUsLinkComponent
Tetrafluoroboric acid diethyl ether complex
Flammable
Corrosive
Tetrafluoroboric acid diethyl ether complex
Catalog # APOSPC450290-100G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Corrosive
Tetrafluoroboric acid diethyl ether complex
Catalog # APOSPC450290-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :161.93 g/mol
Density:1.19 g/cm³ (25 °C)
Flash Pt:129 °C
Storage Temperature:Refrigerator
EINECS:267-983-8
UN:3129
ADR:4.3+8,II
 :GHS02
 :GHS05
CAS Number:67969-82-8
Formula:C₄H₁₁BF₄O
MDL Number:MFCD00011345