Order Entry
Italy
ContactUsLinkComponent
2-(Trimethylsilyl)phenyl trifluoromethanesulphonate
Corrosive
2-(Trimethylsilyl)phenyl trifluoromethanesulphonate
Catalog # APOSPC450251-5G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
2-(Trimethylsilyl)phenyl trifluoromethanesulphonate
Catalog # APOSPC450251-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :298.35 g/mol
Boiling Pt:70 °C/2mmHg
Density:1.229 g/cm³ (25 °C)
Flash Pt:96 °C
Storage Temperature:Refrigerator
UN:3265
ADR:8,II
 :GHS05
CAS Number:88284-48-4
Formula:C₁₀H₁₃F₃O₃SSi
MDL Number:MFCD00799598