
Methyl(2,2,2-trifluoroethyl)amine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2730-52-1
:
Formula:C₃H₇ClF₃N
MDL Number:MFCD08448126
:149.54 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together