[3-(4-Aminophenyl)-5-trifluoromethylpyridin-2-yl]dimethylamine
Catalog # APOSPC446052-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:281.28 g/mol
Storage Temperature:Ambient
CAS Number:1299607-52-5
Formula:C₁₄H₁₄F₃N₃
MDL Number:MFCD18384837