Dimethyl{6-[4-(methylhydrazonomethyl)phenyl]-4-trifluoromethylpyridin-2-yl}amine
Catalog # APOSPC446047-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:322.34 g/mol
Storage Temperature:Ambient
CAS Number:1300019-70-8
Formula:C₁₆H₁₇F₃N₄
MDL Number:MFCD18384856