4-Fluoro-3-(trifluoromethyl)phenylacetic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:220227-47-4
:
Formula:C₉H₆F₄O₂
MDL Number:MFCD00061190
:222.14 g/mol
Melting Pt:50…53 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together