
4-Fluoro-3-(trifluoromethyl)benzonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:67515-59-7
:
Formula:C₈H₃F₄N
MDL Number:MFCD00061284
:189.11 g/mol
Boiling Pt:193…195 °C
Melting Pt:67…69 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS07
Frequently Bought Together