
2-Fluoro-4-(trifluoromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:89763-93-9
:
Formula:C₈H₄F₄O
MDL Number:MFCD00061310
:192.11 g/mol
Boiling Pt:118…119 °C
Density:1.41 g/cm³ (25 °C)
Flash Pt:110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together