
2-Fluoro-6-(2,2,2-trifluoroethoxy)benzenecarbonitrile
Catalog # APOSPC4370B-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:219.14 g/mol
Melting Pt:88…90 °C
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
:GHS06
CAS Number:119584-74-6
Formula:C₉H₅F₄NO
MDL Number:MFCD00068210