
1-(4-Trifluoromethylphenyl)-1,2-propandione 95+%
Catalog # APOSPC4369-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:216.16 g/mol
Boiling Pt:70 °C/0.2mmHg
:GHS07
Storage Temperature:Ambient
CAS Number:10557-13-8
Formula:C₁₀H₇F₃O₂
MDL Number:MFCD01941310