Order Entry
Italy
ContactUsLinkComponent
3,3-Difluorocyclobutane-1,1-dicarboxylic-1-isopropyl ester
Irritant
3,3-Difluorocyclobutane-1,1-dicarboxylic-1-isopropyl ester
Catalog # APOSPC420008-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3,3-Difluorocyclobutane-1,1-dicarboxylic-1-isopropyl ester
Catalog # APOSPC420008-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :222.19 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1225532-90-0
Formula:C₉H₁₂F₂O₄
MDL Number:MFCD23105828