Order Entry
Italy
ContactUsLinkComponent
2-Azaspiro[3.3]heptan-6-ol trifluoroacetate
2-Azaspiro[3.3]heptan-6-ol trifluoroacetate
Catalog # APOSPC420005-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Azaspiro[3.3]heptan-6-ol trifluoroacetate
Catalog # APOSPC420005-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :227.18 g/mol
Storage Temperature:Ambient
CAS Number:1256352-97-2
Formula:C₈H₁₂F₃NO₃
MDL Number:MFCD23703609