2,2,2-Trifluoro-1-(6-(trifluoromethyl)pyridin-3-yl)ethanol 95+%
Catalog # APOSPC409525-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:245.13 g/mol
Storage Temperature:Ambient
CAS Number:1092353-05-3
Formula:C₈H₅F₆NO
MDL Number:MFCD11226776