5-Hydrazino-2-(trifluoromethyl)-1,6-naphthyridine 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:890302-19-9
:
Formula:C₉H₇F₃N₄
MDL Number:MFCD08566918
:228.18 g/mol
Storage Temperature:Ambient
Frequently Bought Together