
4-Isothiocyanato-2-(trifluoromethyl)benzonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:143782-23-4
:
Formula:C₉H₃F₃N₂S
MDL Number:MFCD09800709
:228.19 g/mol
Storage Temperature:Refrigerator
UN:3439
ADR:6.1,III
:GHS07
Frequently Bought Together