Order Entry
Italy
ContactUsLinkComponent
3-Fluoro-5-(pentafluorothio)benzoic acid 98%
Irritant
3-Fluoro-5-(pentafluorothio)benzoic acid 98%
Catalog # APOSPC405678-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Fluoro-5-(pentafluorothio)benzoic acid 98%
Catalog # APOSPC405678-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :266.16 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1448319-13-8
Formula:C₇H₄F₆O₂S
MDL Number:MFCD16652421