Order Entry
Italy
ContactUsLinkComponent
2-Fluoro-4-(pentafluorothio)benzaldehyde 97%
2-Fluoro-4-(pentafluorothio)benzaldehyde 97%
Catalog # APOSPC405671-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Fluoro-4-(pentafluorothio)benzaldehyde 97%
Catalog # APOSPC405671-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :250.16 g/mol
Storage Temperature:Ambient
CAS Number:1800248-21-8
Formula:C₇H₄F₆OS
MDL Number:MFCD16652502