
5-Fluoro-2-nitrobenzotrifluoride 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:393-09-9
:
Formula:C₇H₃F₄NO₂
MDL Number:MFCD00039741
:209.1 g/mol
Boiling Pt:198…199 °C
Melting Pt:23 °C
Density:1.497 g/cm³ (25 °C)
Flash Pt:88 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together