Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Cyano-5-fluorobenzoic acid
3-Cyano-5-fluorobenzoic acid 98%
3-Cyano-5-fluorobenzoic acid 98%
APOSPC3859-5G
:
Apollo Scientific
:
3-Cyano-5-fluorobenzoic acid 98%
APOSPC3859-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
165.12 g/mol
:
171…173 °C
:
Ambient
:
2811
:
6.1,III
:
GHS07
:
327056-74-6
:
C₈H₄FNO₂
:
MFCD05663717