

1,1,1,3,3,3-Hexafluoroisopropyl trifluoromethanesulphonate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:156241-41-7
:
Formula:C₄HF₉O₃S
MDL Number:MFCD02093340
:300.09 g/mol
Boiling Pt:77…78 °C
Density:1.67 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:2927
ADR:6.1+8,II
:GHS05
:GHS06
Frequently Bought Together