


2,2-Difluoroethyl trifluoromethanesulphonate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:74427-22-8
:
Formula:C₃H₃F₅O₃S
MDL Number:MFCD02093342
:214.11 g/mol
Boiling Pt:118…120 °C
UN:2927
ADR:6.1+8,II
:GHS05
:GHS06
:GHS08
Storage Temperature:Ambient
Frequently Bought Together