Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-6-(trifluoromethylthio)benzothiazole
2-Amino-6-(trifluoromethylthio)benzothiazole
2-Amino-6-(trifluoromethylthio)benzothiazole
2-Amino-6-(trifluoromethylthio)benzothiazole
APOSPC3767-100MG
:
Apollo Scientific
:
2-Amino-6-(trifluoromethylthio)benzothiazole
APOSPC3767-100MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 mg
Seq:
2440
:
:
250.27 g/mol
:
GHS07
:
Ambient
:
326-45-4
:
C₈H₅F₃N₂S₂
:
MFCD00665748