Order Entry
Italy
ContactUsLinkComponent
3-(2-Bromophenyl)-5-(2-fluorophenyl)-1,2,4-oxadiazole
3-(2-Bromophenyl)-5-(2-fluorophenyl)-1,2,4-oxadiazole
Catalog # APOSPC3652-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(2-Bromophenyl)-5-(2-fluorophenyl)-1,2,4-oxadiazole
Catalog # APOSPC3652-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:1000339-25-2
Formula:C₁₄H₈BrFN₂O
MDL Number:MFCD09878375