Order Entry
Italy
ContactUsLinkComponent
3-Methyl-5-(trifluoromethoxy)phenylacetic acid 97%
3-Methyl-5-(trifluoromethoxy)phenylacetic acid 97%
Catalog # APOSPC3606-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Methyl-5-(trifluoromethoxy)phenylacetic acid 97%
Catalog # APOSPC3606-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :234.17 g/mol
Melting Pt:52…56 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1000339-57-0
Formula:C₉H₉F₃O₃
MDL Number:MFCD08741405