

α,α,α,2-Tetrafluorotoluene 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:392-85-8
:
Formula:C₇H₄F₄
MDL Number:MFCD00000374
Boiling Pt:114…115 °C/750mmHg
Melting Pt:–49 °C
Density:1.293 g/cm³ (25 °C)
Flash Pt:17 °C
Storage Temperature:Ambient
EINECS:206-880-4
ADR:3,II
:164.1 g/mol
UN:1993
:GHS02
:GHS07
Frequently Bought Together