Order Entry
Italy
ContactUsLinkComponent
2-Fluorobenzenesulphonyl chloride 98%
Corrosive
2-Fluorobenzenesulphonyl chloride 98%
Catalog # APOSPC3488-100G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
2-Fluorobenzenesulphonyl chloride 98%
Catalog # APOSPC3488-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :194.61 g/mol
Boiling Pt:142…146 °C/15mmHg
Melting Pt:26 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
 :GHS05
CAS Number:2905-21-7
Formula:C₆H₄ClFO₂S
MDL Number:MFCD00042285