Order Entry
Italy
ContactUsLinkComponent
1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine 97%
Corrosive
1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine 97%
Catalog # APOSPC3465-5G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine 97%
Catalog # APOSPC3465-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :231.222 g/mol
Boiling Pt:122 °C/2mmHg
Storage Temperature:Ambient
UN:3259
ADR:8,III
 :GHS05
CAS Number:132834-58-3
Formula:C₁₀H₁₂F₃N₃
MDL Number:MFCD00114708