
Ethyl 8-fluoro-4-hydroxyquinoline-3-carboxylate
Catalog # APOSPC3427-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:235.211 g/mol
Melting Pt:217…219 °C
:GHS07
Storage Temperature:Ambient
CAS Number:63010-69-5
:71083-06-2
Formula:C₁₂H₁₀FNO₃
MDL Number:MFCD00052242