Order Entry
Italy
ContactUsLinkComponent
Ethyl-6,7-dichloro-3-(trifluoromethyl)-1,4-dihydroquinoxaline-2-carboxylate
Ethyl-6,7-dichloro-3-(trifluoromethyl)-1,4-dihydroquinoxaline-2-carboxylate
Catalog # APOSPC3358-100MG
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-6,7-dichloro-3-(trifluoromethyl)-1,4-dihydroquinoxaline-2-carboxylate
Catalog # APOSPC3358-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:957062-92-9
Formula:C₁₂H₉Cl₂F₃N₂O₂
MDL Number:MFCD09878367