Order Entry
Italy
ContactUsLinkComponent
1-[3-(Trifluoromethyl)phenyl]but-1-en-3-one 95%
Irritant
1-[3-(Trifluoromethyl)phenyl]but-1-en-3-one 95%
Catalog # APOSPC3344-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-[3-(Trifluoromethyl)phenyl]but-1-en-3-one 95%
Catalog # APOSPC3344-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :214.18 g/mol
Boiling Pt:115 °C/2.5mmHg
 :GHS07
Storage Temperature:Ambient
CAS Number:80992-92-3
Formula:C₁₁H₉F₃O
MDL Number:MFCD00052843