
Ethyl-3-(trifluoromethyl)pyrazole-4-carboxylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:155377-19-8
:
Formula:C₇H₇F₃N₂O₂
MDL Number:MFCD00052083
:208.14 g/mol
Melting Pt:142…148 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together