Ethyl-2-(trifluoromethyl)benzoate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:577-62-8
:31083-13-3
Formula:C₁₀H₉F₃O₂
MDL Number:MFCD00013556
:218.18 g/mol
Boiling Pt:90…91 °C/10mmHg
Density:1.459 g/cm³ (25 °C)
Storage Temperature:Ambient
Frequently Bought Together