


Boron trifluoridedimethyl sulphide complex 95%
Catalog # APOSPC32794-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Cylinder
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:129.94 g/mol
Melting Pt:–16 °C
Density:1.235 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:2924
ADR:3+8,II
:GHS02
:GHS05
:GHS07
CAS Number:353-43-5
Formula:BF₃·S(CH₃)₂
MDL Number:MFCD00013193