Order Entry
Italy
ContactUsLinkComponent
Ethyl-4-hydroxy-8-(trifluoromethoxy)quinoline-3-carboxylate 97%
Irritant
Ethyl-4-hydroxy-8-(trifluoromethoxy)quinoline-3-carboxylate 97%
Catalog # APOSPC3216-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl-4-hydroxy-8-(trifluoromethoxy)quinoline-3-carboxylate 97%
Catalog # APOSPC3216-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :301.2180 g/mol
Melting Pt:235…240 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:175278-35-0
Formula:C₁₃H₁₀F₃NO₄
MDL Number:MFCD00221480