
2-Pyrazin-2-yl-1-(4-trifluoromethylphenyl)ethanol 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:502625-51-6
:
Formula:C₁₃H₁₁F₃N₂O
MDL Number:MFCD03424491
:268.24 g/mol
Melting Pt:100…102 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together