

7H-Dodecafluoroheptanoyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:41405-35-0
:
Formula:C₇HClF₁₂O
MDL Number:MFCD00153143
:364.52 g/mol
Boiling Pt:130…135 °C
Density:1.752 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3265
ADR:8,III
:GHS05
:GHS07
Frequently Bought Together