
1H,1H,7H-Dodecafluoro-1-heptanol 92%
Catalog # APOSPC3130-500G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:332.09 g/mol
Boiling Pt:169…170 °C
Melting Pt:–20 °C
Density:1.762 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:206-406-6
:GHS07
CAS Number:335-99-9
Formula:C₇H₄F₁₂O
MDL Number:MFCD00039630