
2-Benzoxazol-2-yl-1-(4-trifluoromethylphenyl)ethanol 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:502625-50-5
:
Formula:C₁₆H₁₂F₃NO₂
MDL Number:MFCD03424486
:307.27 g/mol
Melting Pt:193…196 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together