
3,5-Bis(trifluoromethyl)benzaldehyde 98%
Catalog # APOSPC3063-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:242.12 g/mol
Boiling Pt:171 °C
Density:1.469 g/cm³ (25 °C)
Flash Pt:70 °C
:GHS07
Storage Temperature:Ambient
CAS Number:401-95-6
Formula:C₉H₄F₆O
MDL Number:MFCD00010206